DURAN® Connector and Syringe Disk Filter, for GL 45 Screw Cap, with 2-Hose Connector, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
DURAN® GL 45 2-hose connection cap for the easy connection of flexible tubing to the DURAN® GL 45 bottles. The two polyproplylene connectors have angled top and straight underside connectors.
- Manufactured from PP, EPDM
- Highly versatile as the screw cap fits the standard GL 45 thread
- 2-port hose connectors allow easy connection of silicone tubing to lab bottles for liquid transfer
- EPDM ensures a gas tight seal with bottle
- Grey polypropylene cap center rotates freely, allowing the bottle to be unscrewed without the need to disconnect the tubing
- Autoclavable at 121 or 134 °C
- Temperature resistant up to +140 °C
- Lab-washer-proof
Grey polypropylene cap center rotates freely, allowing the bottle to be unscrewed without the need to disconnect the tubing. The cap is ideal for use with soft elastic tubing that has an inner diameter of 6 – 9 mm silicone tubing. An optional venting connector with a 0.2 um syringe disk filter is available.
Typical applications: Possible biotech applications include the transfer of sterile media from one container to another using a peristaltic pump.
Customers Who Bought This Also Bought