Standard Opening Crimp Top Vials, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
These vials are designed to work with autosamplers incorporating a robotic arm as well as other autosamplers.
- Suitable for poly Crimp™ seals or standard aluminum seals
- Available with or without graduated marking spots
Vials incorporates the unique step vial design that precisely centers a limited volume insert in the vial neck.
Customers Who Bought This Also Bought