Gaskets, PTFE, Chemglass
Supplier: Chemglass
Certificates
About this item
Gaskets has safety collar to prevent wear as a result of the collar rotating against the stirrer bearing.
- Useful as a dust cover
These PTFE gaskets are 0.015" thick with an opening to fit chemglass stirrer shafts.
Customers Who Bought This Also Bought