A propos de cet article
Designed for histological staining protocols and resin embedding. These thermal transfer printable labels offer excellent chemical resistance and withstand a wide range of temperatures.
- Temperature range: –80 to +149 °C (–112 to +300 °F)
- Able to withstand exposure to harsh chemicals and solvents
- Printable with all major thermal transfer printer brands
- Perfect for labeling microscope slides used throughout the histological staining process
- Resistant to gamma irradiation (tested up to 50 KGy)
- Chemical-resistant labels withstand embedding in histological resins used in microscopy
- Can withstand incubation in temperatures up to 150 °C
- Small size accommodates various sizes of embedding capsules and molds
- Withstands alcohol, xylene, toluene, formalin, as well as histological stains exposure
These labels can be printed with alphanumeric text and 1D/2D barcodes for improved tracking and identification.
These highly glossy labels repel stains and do not acquire colors from histological stains, such as hematoxylin or eosin. The labels and printed information are xylene-resistant when used in combination with the XAR-class of solvent-resistant ribbons. The print will resist xylene, toluene, alcohol, MEK, and other solvents and chemicals without needing additional protection. Ideal for histology lab applications.
Hot resin and chemical-resistant labels for embedding in the resin capsule or mold, alongside the sample. The small size of the label can accommodate various sizes of embedding capsules commonly used in electron microscopy facilities.