Klinger Alpha-Corundum Crystal Model
# de catalogue 470311-034
Fournisseur: Klinger
Spécifications
- Description:Alpha-Corundum Crystal Model
- Cat. no.:470311-034
- Dimensions:36×24×20"
- Weight:8 lbs.
Spécifications
A propos de cet article
This model is hand made in the USA by Klinger Educational Products.
- Contains 135 - 1" balls
This is a permanent structure. We only use grade A materials. The 1" balls are made of hard Maplewood that includes an enameled painted finish. Polished steel rods are used to connect the wooden balls together.
Customers Who Bought This Also Bought