Klinger Aragonite Crystal Model
# de catalogue 470311-032
Fournisseur: Klinger
Spécifications
- Description:Aragonite Crystal Model
- Cat. no.:470311-032
- Dimensions:24×12×12"
- Weight:6 lbs.
Spécifications
A propos de cet article
This model is hand made in the USA by Klinger Educational Products.
- Contains 44 - 1" balls
This is a permanent structure. We only use grade A materials. The 1" balls are made of hard Maplewood that includes an enameled painted finish. Polished steel rods are used to connect the wooden balls together.
Customers Who Bought This Also Bought