Klinger Carbon Collection Crystal Model
# de catalogue 470311-026
Fournisseur: Klinger
Spécifications
- Description:Carbon Collection Crystal Model
- Cat. no.:470311-026
- Dimensions:36×36×12"
- Weight:15 lbs.
Spécifications
A propos de cet article
Set of three Carbon Models, consisting one of each model, Diamond, Graphite and Buckminster Fullerene.
This is a permanent structure. We only use grade A materials. The 1" balls are made of hard Maplewood that includes an enameled painted finish. Polished steel rods are used to connect the wooden balls together.
Customers Who Bought This Also Bought