KIMBLE® HI-VAC® Right-Angle Valve, with Glass Plug and Tip O-Rings, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
This right-angle valve uses a glass plug (as an alternative to PTFE or CTFE) for applications where the 'out-gassing' plug material is undesirable.
- All plugs are totally interchangeable with other HI-VAC® counterparts
- Greater solvent resistance
Manufactured from 33 expansion, low extractable borosilicate glass.
Certifications: Glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Delivery: Supplied with FKM O-rings.
Customers Who Bought This Also Bought