Round Cover Slips, German Glass, Bellco Glass
# de catalogue 76305-514
Fournisseur: Bellco Glass
Spécifications
- Ø:12 mm
- Description:Cover glasses, round, german glass
- Thickness:0.13 - 0.16 mm
- Cat. no.:76305-514
Spécifications
A propos de cet article
These round cover slips are specially developed for fluorescence and confocal microscopy.
- Provides quality reliability and performance
Ideal for neural tissue cell cultures and hard-to-attach.
Packaging: Packed 1 oz. (28 g) per case.
Customers Who Bought This Also Bought