Pasteur Pipette, SUN SRi™
# de catalogue 14235-392
Fournisseur: Thermo Scientific
Spécifications
- Capacity:2 ml
- Length:5³/₄"
- Description:Pasteur pipettes
- Cat. no.:14235-392
- Supplier No.:203 770
Spécifications
A propos de cet article
Pipettes are made of borosilicate glass.
- Straight-cut tips are smooth, fire-polished tops to facilitate bulb attachment
- Without cotton plug
Used in hematology, chemistry, bacteriology as well as general laboratory.
Customers Who Bought This Also Bought