3B Scientific® Stem Of Creeping Buttercup
# de catalogue 470311-856
Fournisseur: American 3B Scientific
Spécifications
- Description:Stem Of Creeping Buttercup
- Height:7 cm
- Cat. no.:470311-856
- Length:28 cm
- Width:28 cm
- No. of Parts:1
Spécifications
A propos de cet article
Cross section of a Creeping Buttercup stem with collateral open vascular bundles. The model shows the typical stem structure of a dicotyledon enlarged by a factor of 250.
Customers Who Bought This Also Bought