3B Scientific® Eye Model
# de catalogue 470311-832
Fournisseur: American 3B Scientific
Spécifications
- Description:Eye Model
- Height:33 cm
- Cat. no.:470311-832
- Length:30 cm
- Width:38 cm
- No. of Parts:12
Spécifications
A propos de cet article
A great model of the anatomy of the human eye.
- Approximately 5x life size
- 58 keyed structures
- Three year manufacturers warranty
- Includes 3B Smart Anatomy
This eye model shows the eyeball with optic nerve in its natural position in the bony orbit (floor and medial wall). Additionally, this eye model shows the relation between eye, bones, muscles, and outer structures of the eye.
BodyTextAccessories: For information about 3B Smart Anatomy, please see the document attached on this page.
Customers Who Bought This Also Bought