Ergomat® Hygiene Cleanroom Anti-Fatigue Food and Medical Floor Mats, Ergomat
Fournisseur: Ergomat
Certificats
A propos de cet article
Hygiene Food and Medical Mat has a broad chemical and pH resistance profile making it perfect for use in areas requiring frequent and caustic sanitation like medical and food processing applications.
- Made of Virgin EPDM rubber
- Can be pressure washed to 1500 psi
- Broad pH resistance from 2 to 13
- Service temperature range −40 to 120 °C
In situations with excessive fluid volume, or the need to promote air flow, Hygiene can be die-cut to accommodate these requirements,
Light Weight and Easy to clean has made this mat a favorite amongst medical professionals as well as in the food industry.
Certifications: ISO Cleanroom Class 5 (Class 100) rated and USDA Certified.
Customers Who Bought This Also Bought