Ellipso-Spoon® Spatula, Chemglass
# de catalogue 89028-940
Fournisseur: Chemglass
Small Business Enterprise
Spécifications
- Length:400 mm
- Material:Stainless steel
- Cat. no.:89028-940
- Supplier No.:CG-1987-04
Spécifications
A propos de cet article
The specialty curved shape permits contact with all corners of bottles, flasks, and containers.
- Made of polished stainless steel
- Handle diameter is 8 mm
Crystals, powders, solid substances and thick liquids are easily retrieved and do not fall back into the container. The sampler has a spoon and spatula.
Customers Who Bought This Also Bought