3B Scientific® Castor Bean Tick
# de catalogue 470311-938
Fournisseur: American 3B Scientific
Spécifications
- Description:Castor Bean Tick
- Height:2 cm
- Cat. no.:470311-938
- Length:12 cm
- Width:12 cm
- No. of Parts:1
Spécifications
A propos de cet article
A common disease spreading tick.
- Large 25:1 scale
- Three years manufacturers warranty
This model of the European sheep tick is a great example of the genus ixodes. The large size allows students to easily examine characteristic tick structure, and is a great accessory when discussing tick born diseases like Lyme disease.
Customers Who Bought This Also Bought