ContactUsLinkComponent
3B Scientific® Dual Sex Torso with Asian Features
3B Scientific® Dual Sex Torso with Asian Features
# de catalogue 470311-816
Fournisseur:  American 3B Scientific
Model Deluxe Asian Dual Sex Torso 18Part
3B Scientific® Dual Sex Torso with Asian Features
# de catalogue 470311-816
Fournisseur:  American 3B Scientific
Spécifications
  • Description:
    Asian Dual-Sex Torso
  • Height:
    85 cm
  • Cat. no.:
    470311-816
  • Length:
    15 cm
  • Width:
    24.9 cm
  • No. of Parts:
    18
Spécifications
A propos de cet article
Developed for schools in asia.
  • Includes 3b torso guide
  • 317 structures keyed from photos
  • Three year manufacturers warranty
  • Includes 3B Smart Anatomy
The 3B Scientific Torso offers many possibilities you need for detailed demonstrations in human anatomy. With this human torso model you receive 100% quality and a high standard of detailed manufacturing. The quality of this torso is impressive, just like the price! Use it to answer all questions on internal human anatomy. This model of a human torso was developed especially for Asian schools. The internal organs are soft, durable plastic and can be removed.
This dual-sex torso is complete as a tool for human anatomy education with the 3B Torso Guide. All the organs in this human torso are hand painted for a quality product. This great human anatomy educational tool and makes learning the location of the human organs easy.
BodyTextAccessories: For information about 3B Smart Anatomy, please see the document attached on this page.
Customers Who Bought This Also Bought