3B Scientific® Physical Eye Model
# de catalogue 470311-948
Fournisseur: American 3B Scientific
Spécifications
- Description:Physical Eye Model
- Cat. no.:470311-948
Spécifications
A propos de cet article
How does an eye work?
- Examine eye function and the interaction of lenses
- Identify common problems like near and far sightedness
- Includes case for storage or transport
This physical eye model can be used to demonstrate the optical functions of the human eye, e.g. representation of an object on the retina, accommodation (change in the lens curvature), short-sightedness and far-sightedness.
Customers Who Bought This Also Bought