3B Scientific® Fetal Development Models
Fournisseur: American 3B Scientific
Certificats
A propos de cet article
Individual models showing fetal development.
- Individual models on stands
- Available individually or as a set
- Three years manufacturers warranty
- Includes 3B Smart Anatomy
Examine fetal development from four weeks to seven months with these detailed models from 3B Scientific. For consistency each model (1 - 8) utilizes the same identified structures, allowing students to observe the changes that occur during development.
The final model (four week embryo) is labeled with structures specific to the embryo. As a set these make a great addition to any biology, development, or human reproduction curriculum.
BodyTextAccessories: For information about 3B Smart Anatomy, please see the document attached on this page.
Customers Who Bought This Also Bought