55 Gallon Open-Head UN Rated Steel Drum, New Pig®
Fournisseur: New Pig
Certificats
A propos de cet article
Tough, UN Rated drums are made for hazardous materials; you can rely on them to deliver safety and compliance.
- UN Rated and tested to help ensure safe shipping of hazardous materials
- Open-head design and removable lid offer full, convenient access to inside of drum
- 12-gauge bolt ring closure, sponge rubber gasket and plated bolt/nut provide tight seal to help prevent leaks and spills
- Interior is coated with a corrosion inhibitor; choose optional phenolic/epoxy lining for added resistance
16-gauge steel lid and 18-gauge steel body and bottom meet strict wall thickness requirements to ensure strength, durability and long life.
Customers Who Bought This Also Bought