BradyPrinter i5100 Industrial Label Printer, Brady®
Fournisseur: Brady Worldwide
Certificats
A propos de cet article
The BradyPrinter i5100 industrial label printer provides the fundamental printing capabilities for the high-mix and high-volume work you need to get done every day.
- Single color printing
- 5000 labels per day (8.5 hours)
- Intuitive touchscreen to easily change settings
- Easy lock design for changing and placing materials
- Interchangable rollers
- Manual or auto-cutter options
- Five year warranty
Smart printing technology allows the materials, printer and software to communicate, making set-up faster and taking the guess work out of material changeovers. Specially designed ribbon spindles guide and roll holders that lock into place without any clumsy extra steps. Printer offers a variety of connection ports to stay connected any way you need.
Certifications: Unit ships with Brady Workstation Product and Wire ID Software Suite with built-in templates for fixed text or free-form label design options.
Ordering information: Includes one RFID-enables media roll holder, one ribbon takeup core, power cord, USB cable, user license for software suite, CD that contains drivers, and user maual.
Spécifications
- Catalog No:
- 76305-444
- 76305-446
- 76305-442
- 76305-440
- Connections:
- Bluetooth, Ethernet, Serial RS232, USB 2.0, USB Host
- Bluetooth, Ethernet, Serial RS232, USB 2.0, USB Host
- Bluetooth, Ethernet, Serial RS232, USB 2.0, USB Host
- Bluetooth, Ethernet, Serial RS232, USB 2.0, USB Host
- Display:
- Color Touch Screen, 4.3" (diagonal)
- Color Touch Screen, 4.3" (diagonal)
- Color Touch Screen, 4.3" (diagonal)
- Color Touch Screen, 4.3" (diagonal)
- Electrical:
- 100/240 V, 50/60 Hz
- 100/240 V, 50/60 Hz
- 100/240 V, 50/60 Hz
- 100/240 V, 50/60 Hz
- Label size:
- 6.60' max./0.20" min. length
- 6.60' max./0.20" min. length
- 6.60' max./0.20" min. length
- 6.60' max./0.20" min. length
- Model:
- i5100 Standard Version
- i5100 Auto Cutter Version
- i5100 Auto Cutter Version
- i5100 Standard Version
- Dimensions:
- 12.5H×9.5W×17"D
- 12.5H×9.5W×17"D
- 12.5H×9.5W×17"D
- 12.5H×9.5W×17"D
- Weight:
- 13.6 lbs.
- 14.6 lbs.
- 14.6 lbs.
- 13.6 lbs.