Safety Vest, Solid Front with Mesh Back, Class II, ML Kishigo
Fournisseur: M.L. KISHIGO
Certificats
A propos de cet article
Class II high visibility safety vest, provides enhanced visibility to people working in hazardous situations under all light conditions.
- Zipper front closure
- Left and right chest mic tabs
- Two lower outside patch pockets with flaps
- Right chest two-tier, four-division pocket
- Left chest radio pocket with flap
- Two lower inside patch pockets
Back of vest is made of a comfortable Ultra-Cool™ 100% polyester mesh material. To enhance visibility the vest offers a 2" wide high performance reflective material with 3" contrasting color.
Certifications: ANSI 107 Type R. Class 2 Compliant.
Spécifications
- Catalog No:
- 76203-662
- 76203-660
- 76203-586
- 76203-666
- 76203-664
- 76203-654
- 76203-596
- 76203-652
- 76203-598
- 76203-658
- 76203-648
- 76203-656
- 76203-588
- 76203-590
- 76203-650
- 76203-594
- Garment application:
- Industrial, Outdoor, Services
- Industrial, Outdoor, Services
- Industrial, Outdoor, Services
- Industrial, Outdoor, Services
- Industrial, Outdoor, Services
- Industrial, Outdoor, Services
- Industrial, Outdoor, Services
- Industrial, Outdoor, Services
- Industrial, Outdoor, Services
- Industrial, Outdoor, Services
- Industrial, Outdoor, Services
- Industrial, Outdoor, Services
- Industrial, Outdoor, Services
- Industrial, Outdoor, Services
- Industrial, Outdoor, Services
- Industrial, Outdoor, Services
- Risk area:
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- HiVis
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Closure type:
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Fabric:
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester