93-360 Thinnest, Chemical Resistant Disposable Cleanroom Gloves, Microflex®, Ansell
Fournisseur: Cardinal Health
Certificats
A propos de cet article
This is made of an innovative, synthetic composite material for tough chemical protection.
- Three layer design for superior protection
- Thin construction
- Easy donning
- Non-particulating packaging
The extra soft formulation and ergonomic design provide a comfortable fit and feel for extended wear times.
Certifications: CE 0086 marked. EN ISO 374-1 Type A: JKLOPST chemical hazards. EN ISO 374-5: VIRUS. EN 388: 2000X mechanical hazards. EAC TP to TC 0192011. EN 421.
Spécifications
- Catalog No:
- 76289-852
- 76289-850
- 76289-856
- 76289-854
- 76289-860
- 76289-858
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Material:
- Neoprene/Nitrile
- Neoprene/Nitrile
- Neoprene/Nitrile
- Neoprene/Nitrile
- Neoprene/Nitrile
- Neoprene/Nitrile
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Anti-static:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes