Cryokit 550, 22" Cryogenic Gloves, Mapa Professional
Fournisseur: MAPA
Certificats
A propos de cet article
These gloves are designed for applications handling liquid nitrogen and other cryogenic gases, to protect from cold contact and prevent burns from liquid gas leakage.
- Multilayer fleece lining
- Ventilation technology
- Waterproof
Gloves remain ventilated due to the inner polyolefin porelle membrane which allows for air to flow and keeps hands dry.
A special elastic laminated blue fabric provides dexterity and comfort in the glove and cuff areas.
Certifications: CE Cat. III 0498 marked. Cold Hazards EN511: 231. Mechanical Hazards EN388: 2422.
Spécifications
- Catalog No:
- 75841-236
- 75841-234
- 75841-232
- 75841-230
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Polyolefin
- Polyolefin
- Polyolefin
- Polyolefin
- Material:
- Polyester
- Polyester
- Polyester
- Polyester
- Finish:
- Elastic Laminated Blue Fabric
- Elastic Laminated Blue Fabric
- Elastic Laminated Blue Fabric
- Elastic Laminated Blue Fabric
- Cuff style:
- Gauntlet
- Gauntlet
- Gauntlet
- Gauntlet