A propos de cet article
Industry's first high-speed centrifuge and 3 l magnetic stirrer combined into one unit. Enables a substantial cost savings for labs by not having to purchase two pieces of equipment, a stirrer and a centrifuge, because the MagFuge® does it all.
- Modern low profile design is compact and has a small foot print
- Easy to use back-lit digital panel
- Programable digital control
- Rotor sensing technology
- Long-life maintence free brush-less motor
- Five-year warranty
- Power source: Universal power cord
The MagFuge® is the industry’s first high-speed centrifuge (12,500 rpm) and magnetic stirrer (3 l), combined into one unit. The design innovation enables a substantial cost savings for labs by not having to purchase two individual pieces of equipment, a stirrer and a centrifuge, because the MagFuge® does it all.
The high-speed centrifuge is designed to give you maximum speed and flexibility and is comfortable and easy to use. Safety features ensure it will not operate when the lid is open, a rotor is jammed, or the unit is out of balance. Ideal for most protocols requiring fast spins (12,500 rpm/9,800xg), and appropriate for PCR, microfilter cell separation, and HPLC protocols.
The magnetic stirrer offers a powerful magnetic coupling that ensures consistent accurate alignment of the stir bar that significantly reduce the chance of spin-outs. After hours of continuous use, the unit remains cool ensuring sample integrity. A silicone mat is included to help prevent spills and to help keep vessels stable.