Klinger Carbon Dioxide Crystal Model
# de catalogue 470311-014
Fournisseur: Klinger
Spécifications
- Description:Carbon Dioxide Crystal Model
- Cat. no.:470311-014
- Dimensions:24×24×12"
- Weight:8 lbs.
Spécifications
A propos de cet article
This model is hand made in the USA by Klinger educational products.
- Permanent structures
- Made with grade A materials
The 1" balls are made of hard Maplewood that includes an enameled painted finish. Polished steel rods are used to connect the wooden balls together.
Customers Who Bought This Also Bought