xPIPETTE™ Adjustable Volume Ergonomic Pipettes, Single and Multichannel, Biotix
Supplier: Biotix
Certificates
About this item
xPIPETTES™ offer accuracy and precision with low ejection forces. Biotix uses patented tip ejection technology to reduce fatigue and makes the xPIPETTE a top choice to improve your lab’s ergonomics.
- Fully compatible with Rainin LTS tips and Biotix xTIP4™ tips
- Calibrated to ISO 8655 standards with Biotix xTIP4 tips
- Light spring forces produce a smooth and controlled aspiration with perfect balance
- Padded ejector button to reduce hand vibrations during tip ejection
- Simple volume adjustment with a secure lock to prevent volume drift
- Available in single, 8, and 12-channel models with a symmetrical design for ambidextrous use
These pipettes are designed with durable and lightweight components. They deliver a precise and accurate liquid transfer while maintaining optimal hand comfort.
Certifications: ISO 8655 certified. Calibration certificate included in the box with each pipette. Unique serial ID number under the finger hook for GLP/GMP compliance.
Caution: Rainin and LTS are a trademark of Mettler-Toledo, Rainin LLC.
Customers Who Bought This Also Bought