Whatman™ Application-Specific Filter Papers, Grade 287 ½, Whatman products (Cytiva)
Fournisseur: Cytiva
Certificats
A propos de cet article
Kieselguhr paper with a medium to slow flow rate. Additional adsorption effect, e.g., for the separation of very fine semi-colloidal turbidity, for clarifying milk serum, starch solutions, soil suspensions or sugar-containing solutions prior to polarimetry or refractometry.
Customers Who Bought This Also Bought