Aurelia® Quest 2.2® Flexible Nitrile Powder-Free Examination Gloves, Supermax
Fournisseur: SUPERMAX HEALTHCARE CANADA CA
Certificats
A propos de cet article
Aurelia® Quest 2.2® gives the wearer even greater tactile sensitivity, comfort and flexibility while keeping the fit and look of latex. Ultra-sheer yet durable.
- 100% latex-free
- Food contact approved
- Finger-textured
- Beaded cuff for ease of donning
Certifications: CE marked. AQL 1.5. EN 455 CE Class 1 Medical Grade. EN 420, EN 388 & EN 374. EN 1186 Food Contact Approved.
Spécifications
- Catalog No:
- CA76093-785
- Ambidextrous/Hand-specific:
- Ambidextrous
- Disposable / reusable:
- Disposable
- Latex/Latex-free:
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Cuff style:
- Beaded
- Sterility:
- Non sterile
- Food contact approved:
- Yes