About this item
Microbiological incubator series has been updated for optimum performance. Ranging in sizes from 57 to 331 liters (2 to 11.7 cu.ft.), the SMI series is guaranteed to fit any workspace. Ideal for clinical, analytical, and general laboratory applications.
- USB data logging capabilities
- Independent over temperature set point and operational control override for additional safety
- Side-mounted access port, 38 mm diameter (1.5")
- Stainless steel interior construction for long life operation and easy cleaning
- Condensed connection panel includes; 4 to 20 mA analog output, dry contact port, and USB A port
- Available in 100 to 120 V or 220 to 240 V configurations
- 24 month limited warranty
- Perfect solution for general cell culture and constant temperature applications
Heated external doors prevent cold spots and maintain thermal uniformity. Unique air-jacketed design provides excellent temperature uniformity of ±0.5 °C at 37 °C. Interior chamber outlet (1 amp). Sealed glass inner door allows for viewing without disturbing the critical growth atmosphere.
Heating rely on natural heat radiation for heating. Over Temperature Limit system (OTL) is a mechanical heating cutoff included with each incubator and operates independently of the incubator microprocessor temperature controller, this helps safeguard the incubator by preventing runaway heating in the event of hardware failures or a sudden external heat spike.
Capacitive touch screen of 7" usable with laboratory gloves. With on-screen graphing and real time data, the new SMI controller provides the highest levels of sample security and traceability. Touch screen controller features: Storage up to 5 years of log data, password locking capabilities, data logged once per minute, export in .CSV file, smart alarm (accounts for door openings and temperature set point changes before alarm), temperature deviation alarm.