Laboratory Installation SafetyWasteCaps, Collector for HPLC Waste, S.C.A.T
Fournisseur: S.C.A.T.
Certificats
A propos de cet article
SafetyWasteCaps conduct the waste tubes of your HPLC system to the disposal system, directly and air-tight. Escape of harmful solvent vapours are effectively prevented.
All parts coming in contact with fluids are manufactured from pure PTFE - for optimum chemical resistance even against aggressive solvents and acids.
The useful 3-way and 4-way collectors both can be used as extraction as well as waste management connection to your HPLC systems. They fit to GL 45 bottles and the laboratory installation tube system.
Customers Who Bought This Also Bought