ContactUsLinkComponent
Oct4 Transcription Factor Assay Kit, Cayman Chemical Company
Oct4 Transcription Factor Assay Kit, Cayman Chemical Company
# de catalogue 75817-098
Numéro CAS:  
Oct4 Transcription Factor Assay Kit
Oct4 Transcription Factor Assay Kit, Cayman Chemical Company
# de catalogue 75817-098
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Oct4 Transcription Factor Assay Kit, 96 Strip Wells
  • Cat. no.:
    75817-098
  • Supplier No.:
    601080-96
Spécifications
A propos de cet article
96-Well assay for measurement of Oct4.
  • A sensitive, non-radioactive method of detecting Oct4 from whole cell lysates
  • 96-well plate format replaces EMSAs
  • Capture the transcription factor using a specific dsDNA sequence bound to the plate
  • Detect the dsDNA-bound transcription factor with specific antibodies in an ELISA format
  • Nuclear extraction kit is available to aid in the isolation of nuclear and cytoplasmic fractions from cell lysates or tissue homogenates
Octamer-binding transcription factor 4 (Oct4) is an essential regulator of embryonic stem cell pluripotency and used as a marker for undifferentiated cells. It also plays a role in gene expression, DNA repair and/or replication, signal transduction and tumorigenesis. Cayman’s Oct4 Transcription Factor Assay is a non-radioactive, sensitive method for detecting specific transcription factor-DNA binding activity in nuclear extracts. A 96-well enzyme-linked immunosorbent assay (ELISA) replaces the cumbersome radioactive electrophoretic mobility shift assay (EMSA). A specific double-stranded DNA (dsDNA) sequence containing the Oct4 response element is immobilized onto the wells of a 96-well plate. Oct4, contained in a nuclear extract, binds specifically to the Oct4 response element. Oct4 is detected by the addition of a specific primary antibody directed against Oct4. A secondary antibody conjugated to HRP is added to provide a sensitive colorimetric readout at 450 nm.
Delivery: Kit contains assay buffer, reagent A, positive control, binding buffer, primary antibody, wash buffer concentrate, polysorbate 20, competitor dsDNA, HRP conjugate, plate and cover sheet, developing and stop solution.
Customers Who Bought This Also Bought