Sludge Nabber, Nasco Sampling
Fournisseur: Avantor
Certificats
A propos de cet article
Sample bottles of various sizes are held via an adjustable locking device.
- Makes sample collection of liquids fast and easy
- Flexible with adjustable aluminum pole
- PU holder with a plastic snapper ring
Suitable for numerous requirements including chlorine in solution – Can also be used for BOD.
Delivery: Supplied with a 500 ml PE bottle.
Customers Who Bought This Also Bought