Sparkle Labs E-Textile Kit
# de catalogue 470311-254
Fournisseur: SPARKLE LABS
Spécifications
- Description:Sparkle Labs E-Textile Kit
- Cat. no.:470311-254
Spécifications
A propos de cet article
The E-Textile Lab Pack lets you explore the wonderful world of electronic sewing (e-sewing) and e-textiles through a series of introductory projects using the system. You will learn how to sew basic circuits to light up LEDs.
- LEDs are fun forms and unique colors
- Easy to sew light up e-textile project
- Up to ten projects can be made with the kit
- Microcontroller-compatible
Illustrated step-by-step instructions for using your pieces to create a sewable circuit project with conductive thread. Easy-to-follow diagrams and troubleshooting tips make this a great introductory resource for crafters and creatives.
Delivery: Includes 10×RGB Moon LED; 10×Coin Cell Battery Holder - switched - 20 mm; 10×Needle Set; 10×Conductive Thread Bobbin - 30' (stainless steel); 10×Coin Cell Battery - 20 mm (CR2032); 2×Star LED - Pink (5 pcs); 2×Heart LED - Red (5 pcs); 2×Small round LED - Aqua (5 pcs); 2×Small round LED - Yellow (5 pcs); 4×Small round LED - Orange (5 pcs).
Customers Who Bought This Also Bought