Rainbow LED Pack
# de catalogue 470311-296
Fournisseur: SPARKLE LABS
Spécifications
- Description:Rainbow LED
- Cat. no.:470311-296
Spécifications
A propos de cet article
A rainbow of colors in nine clear 5 mm LEDs and one white 10 mm LED. These are standard, super-bright LEDs. You can use them with a 3 V power supply or in series with a resistor and a 5 V power supply.
- Clear LEDs that come in lime, blue, pink, purple, yellow, aqua, orange, red, green and white
- Breadboard ready
- Beautiful colors
Nine colors included plus one bonus 10 mm LED: red, orange, yellow, lime, green, blue, aqua, violet, white. Let us make some rainbows. 5 mm LEDs. Great for breadboards too.
Customers Who Bought This Also Bought