VWR® Upholstered Lab Stools
Fournisseur: VWR International
Certificats
A propos de cet article
This VWR® upholstered lab stool features a roomy and comfortable 16" Ø seat with 3.5" of resilient foam cushioning and adjustability proven to provide reliable ergonomic performance, shift after shift, year after year.
- Exclusive black microdot-textured vinyl upholstery combines practical performance with a sophisticated look
- Other components include an easy-to-use ergonomic task control with seat height and back tilt adjustments and 26" Ø black composite base
- Designed for laboratory applications, including those in healthcare, education and industrial settings
- Backed by a 10-year warranty (5 years on upholstery, lifetime on pneumatic)
The VWR® lab chair line features a refined yet pragmatic collection of chairs and stool designed for today’s laboratory professionals, applications and settings. The combination of european design with proven ergonomic functionality and engineering results in a focused set of models that meet real-world lab seating challenges.
Certifications: Meets ANSI/BIFMA standards for durability and safety.
Caution: This product can expose you to chemicals including wood dust, which is known to the State of California to cause cancer. For more information go to www.p65warnings.ca.gov.
Spécifications
- Catalog No:
- 76308-780
- 76308-784
- 76308-788
- 76308-792
- Model:
- 76308-780
- 76308-784
- 76308-788
- 76308-792
- For use with:
- Medium height benches (34.75-44")
- Medium height benches (34.75-44")
- Low benches (29-34")
- Low benches (29-34")
- Seat/back color:
- Black
- Black
- Black
- Black
- Seat/back material:
- Vinyl
- Vinyl
- Vinyl
- Vinyl
- Seat dimensions:
- Ø16×T3.5"
- Ø16×T3.5"
- Ø16×T3.5"
- Ø16×T3.5"