VWR® Lab Chairs, PP
Supplier: VWR International
Certificates
About this item
This durable VWR® polypropylene lab chair features antimicrobial properties and UV inhibitors integrated in its seat and backrest, and the ergonomic functionality and versatility to make it an ideal choice for a variety of laboratory types and applications.
- Rugged, easy-to-clean design highlighted by black polypropylene seat and back with antimicrobial properties and UV inhibitors
- Other components include an easy-to-use ergonomic task control with seat height and back tilt adjustments and 26" diameter black composite base
- Designed for laboratory applications, including those in healthcare, education and industrial settings
- Backed by a 10-year warranty (lifetime on pneumatic)
The VWR® Lab Chair Line features a refined yet pragmatic collection of chairs and stool designed for today’s laboratory professionals, applications and settings. The combination of european design with proven ergonomic functionality and engineering results in a focused set of models that meet real-world lab seating challenges.
Certifications: Meets ANSI/BIFMA standards for durability and safety.
Caution: This product can expose you to chemicals including wood dust, which is known to the State of California to cause cancer. For more information go to www.p65warnings.ca.gov.
Specifications
- Catalog No:
- 76308-756
- 76308-760
- 76308-758
- 76308-762
- Model:
- 76308-756
- 76308-760
- 76308-758
- 76308-762
- For use with:
- Medium height benches (32.75 - 42")
- Low benches (27-32")
- Medium height benches (32.75 - 42")
- Low benches (27-32")
- Frame material:
- Composite
- Composite
- Composite
- Composite
- Seat/back color:
- Black
- Black
- Black
- Black
- Seat/back material:
- PP
- PP
- PP
- PP
- Seat dimensions:
- W17.5X×D15.5
- W17.5X×D15.5
- W17.5X×D15.5
- W17.5X×D15.5
- Backrest dimensions:
- W16×H10.75"
- W16×H10.75"
- W16×H10.75"
- W16×H10.75"