ContactUsLinkComponent
Sewable LED Moons
Sewable LED Moons
Catalog # 470311-288
Supplier:  SPARKLE LABS
SEWABLE MOONS PACK 2
Sewable LED Moons
Catalog # 470311-288
Supplier:  SPARKLE LABS
Specifications
  • Description:
    Sewable LED Moons
  • Cat. no.:
    470311-288
Specifications
About this item
Color changeable sewable moons. Add cool decorations and color to your interactive textile projects. These moons can be used without a microcontroller but requires a sewable battery holder. Use it with your LilyPad or other textile ready microcontroller.
  • RGB programmable colors
  • Fun design to go along with the sewable stars
  • Resistors are on the board
  • Larger size so they can be easily sewn
These cool LEDs have a built-in resistor. They rely on conductive thread. Our high-quality conductive thread is a perfect companion to these sewable moons. With the battery removed, fabrics with electronics can be washed. Conceptually, these circuits are identical to their wire-and-solder counterparts, observing the same principles of polarity and resistance. Only the materials, methods and sometimes scale are different. There is a lot more to sewable circuits than LEDs. Check out the Sparkle Labs E-Textile kit.
Customers Who Bought This Also Bought