Containers, CTH Series, Eagle Stainless
Supplier: Eagle Stainless Container
Certificates
About this item
Containers, CTH series with clip down type closure.
- Made of 316L stainless steel container and with lid
- Electropolished finish (10 Ra)
- With passivation option
Seamless sanitary interior and exterior with radiused bottom-corners and has 304 stainless steel clips with hollow side handles.
Air/water-tight seal with platinum-cured silicone gasket (Class VI Certification CTH-18 through CTH-47H).
Customers Who Bought This Also Bought