Caps, GL Thread with Aperature, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
Cap is made from PBT (thermoplastic polyester). It can be combined with the sealing ring will accommodate tubing of the listed diameter.
- Open top cap for use with GL screw threads
- Can withstand temperatures from –45 to 200 °C
Customers Who Bought This Also Bought