A propos de cet article
A competitive assay that can be used for quantification of LTB4 in plasma and other sample matrices.
- Assay 24 samples in triplicate or 36 samples in duplicate
- Measure LTB4 levels down to 45 pg/ml
- Rapid assay; get results in under five hours
LTB4 stimulates a number of leukocyte functions, including aggregation, stimulation of ion fluxes, enhancement of lysosomal enzyme release, superoxide anion production, chemotaxis, and chemokinesis.
LTB4 is metabolized in leukocytes and hepatocytes to less active 20-hydroxy and 20-carboxy LTB4 and then further oxidized to ω-carboxy dinor LTB4 and ω-carboxy tetranor-LTB₃. It is not excreted in the urine.