Order Entry
ContactUsLinkComponent
PAD4 Autoantibody ELISA Kit, Cayman Chemical Company
PAD4 Autoantibody ELISA Kit, Cayman Chemical Company
# de catalogue 75816-822
Numéro CAS:  
PAD4 Autoantibody ELISA Kit
PAD4 Autoantibody ELISA Kit, Cayman Chemical Company
# de catalogue 75816-822
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Detection range:
    15.6 - 1000 U/ml
  • Storage temperature:
    4 °C
  • Cat. no.:
    75816-822
  • Supplier No.:
    500930-96
  • No. of Tests:
    96 Wells

Spécifications

A propos de cet article

An immunometric for the measurement of anti-PAD4 autoantibodies.

  • Measure anti-PAD4 autoantibodies of any isotype (IgM, IgG, IgA)
  • For use in human plasma and serum (no need to purify samples)
  • Lower Limit of quantification is 31.3 U/ml

PAD4 is a guanidino-modifying enzyme which catalyzes the conversion of specific arginine residues to citrulline in a calcium-dependent manner. PAD4 activity is increased in rheumatoid arthritis (RA), producing citrulline-containing proteins (CCP) that induce the formation of arthritogenic autoantibodies.

In addition to anti-CCP autoantibodies, 23 to 45% of RA patients also produce autoantibodies specific for the PAD4 enzyme itself. PAD4 Autoantibody ELISA Kit is an immunometric assay which can be used to measure anti-PAD4 autoantibodies of any isotype (IgM, IgG, and IgA) in human plasma and serum without prior sample purification.

Affinity-purified anti-PAD4 antibody isolated from the plasma of a patient with RA is used as the standard. One unit is approximately equal to 1 ng of anti-PAD4 Ig protein.

Delivery: Kit contains HRP conjugate, plate, ELISA standard, wash buffer concentrate, non-fat dry milk assay reagent, ELISA buffer concentrate, polysorbate 20, substrate and stop solution, cover sheet.
Customers Who Bought This Also Bought