Order Entry
ContactUsLinkComponent
PPARα Transcription Factor Assay Kit, Cayman Chemical Company
PPARα Transcription Factor Assay Kit, Cayman Chemical Company
# de catalogue 75816-696
Numéro CAS:  
PPARA Transcription Factor Assay Kit
PPARα Transcription Factor Assay Kit, Cayman Chemical Company
# de catalogue 75816-696
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Storage temperature:
    −80 °C
  • Cat. no.:
    75816-696
  • Supplier No.:
    10006915-96
  • No. of Tests:
    96 Wells

Spécifications

A propos de cet article

A 96-well assay for measurement of PPARα DNA binding activity.

  • A sensitive, non-radioactive method of detecting PPARα from whole cell lysates
  • 96-well plate format replaces EMSAs
  • Capture the transcription factor using a specific dsDNA sequence bound to the plate
  • Detect the dsDNA-bound transcription factor with specific antibodies in an ELISA format

PPARs are ligand-activated transcription factors belonging to the large superfamily of nuclear receptors. PPARα primarily activates genes encoding proteins involved in fatty acid oxidation, while PPARγ primarily activates genes directly involved in lipogenic pathway and insulin signaling.

Members of the PPAR family are important direct targets of many antidiabetic and hypolipidemic drugs. PPAR Transcription Factor Assays are a non-radioactive, sensitive method for detecting specific transcription factor DNA binding activity in nuclear extracts and whole cell lysates.

Delivery: Kit contains assay buffer, Reagent A, positive control, binding buffer, antibody, concentrate, polysorbate 20, competitor dsDNA, HRP conjugate, plate and cover, developing and stop solution.
Customers Who Bought This Also Bought