A propos de cet article
A competitive assay that permits the rapid measurement of PGD2 from biological samples.
- Treatment of samples with MOX hydrochloride converts unstable PGD2 to stable PGD2-MOX, MOX derivatization reagent included with assay
- Assay 24 samples in triplicate or 36 samples in duplicate
- Measure PGD2-MOX levels down to 16 pg/ml
- Rapid assay; get results in under four hours
PGD2-MOX Express ELISA is a competitive assay that permits the rapid measurement of PGD2 from biological samples, requiring only one-hour incubation and development times for each step. The methodology for the assay is the same as that employed in the PGD2-MOX ELISA (Item No. 512011). The ELISA offers the convenience of a fast assay while still achieving a sensitivity of ~16 pg/ml and an assay range of 7.8 to 1000 pg/ml.