Chains for Gas Cylinder Support Brackets, Justrite®
Fournisseur: Justrite
Certificats
A propos de cet article
These chains provide safety when moving and storing of gas cylinders.
- Chains are 1041 mm (41") long
For an improved level of safety for any installation, most brackets, stands, and carts include openings to retrofit secondary support chains, one chain for each cylinder strap.
Welded and plated, heavy-duty size 2/0 steel chains with twist link, or Type 316 stainless steel 5 mm (0.187") proof coil chain allows links to lay flat against the cylinders, enabling a tighter fit and preventing damage to required labeling.
Customers Who Bought This Also Bought