One - Hour Fire Barrier, Justrite®
Fournisseur: Justrite
Certificats
A propos de cet article
One-Hour Fire Barrier provides safety when moving and storing of gas cylinders.
- One hour fire wall rating
- Each barrier includes six 0.0406" diameter mount holes
Constructed using fire-resistant Marinite® I fully encased into welded steel panels, these units may be mounted to exterior walls and floor using provided mounting holes with self-supplied fasteners, or they may be used as a free-standing wall.
Lift hook plates supplied for installation positioning.
Customers Who Bought This Also Bought