Medical Oxygen Cylinder Holder, Justrite®
Fournisseur: Justrite
Certificats
A propos de cet article
This Medical Oxygen Cylinder Holder provides safety when moving and storing of gas cylinders.
- Constructed using steel with corrosion-resistant powder coating
- Stands include 0.312" diameter mounting holes for permanent installations
- Used for medical oxygen or calibration gases in laboratories
This ring stand uses 10 mm (0.375")nylon set screws to hold cylinders in place.
Certifications: Holders are NFPA 1 and NFPA 55 approved, OSHA approved, and CGA approved.
Spécifications
- Catalog No:
- 76292-550
- 76292-554
- 76292-552
- Cylinder type:
- Compressed gas
- Compressed gas
- Compressed gas
- Material:
- Steel
- Steel
- Steel
- Mounting type:
- Bench
- Bench
- Bench