Seismic-Rated Anchor for Barricade Racks, Justrite®
# de catalogue 76292-556
Fournisseur: Justrite
Spécifications
- Depth:½"
- Width:½"
- Description:Accessories for gas cylinder equipment
- Height:5½"
- Material:Steel
- Cat. no.:76292-556
- Weight:0.3 lbs.
Spécifications
A propos de cet article
This Seismic-Rated Anchor for Barricade Racks provides safety when moving and storing of gas cylinders.
- Listed for use in seismic zones A, B, C, D, E and F
- For use with barricade style racks
Comprised of a high-strength threaded stud body, expansion clip, nut and washer, these seismic wedge-type anchors are designed for unbroken concrete.
Bodies are made of plated carbon steel, while the expansion clip consists of a split cylindrical ring with undercutting grooves.
Certifications: 2015 IBC Compliant and anchors are tested to ACI 355.2 and ICC-ES AC193.
Customers Who Bought This Also Bought