Gas Cylinder Process Stand, Justrite®
Fournisseur: Justrite
Certificats
A propos de cet article
This Gas Cylinder Process Stand provides safety when moving and storing of gas cylinders.
- Securely mount cylinder controls and cross-over devices with integral elevated panels
- Welded construction using 11 gauge (3 mm) steel provides structural integrity and longevity
- Exclusive bumper guard edging with steel reinforced vinyl protects cylinders from damage while providing extra grip
- Accepts cylinders from 102 to 305 mm (4 to 12") in diameter
- Prevent dangerous tipping situations
- Safe storage and distribution of industrial and commercial use compressed gas cylinders
- Powder-coated to ensure long service life and chemical resistance
- Back-to-back cylinder storage models allow for maximum space utilization
Pre-drilled holes in base for floor mounting with application specific fasteners (not included).
Two heavy-duty polypropylene support straps, 38×1372 mm (1.5×54") and rated a t516 kg (1138 lbs.), feature steel cinch buckle and hold individual cylinders securely against integral bracket.
For an improved level of safety for any installation, all brackets include openings to retrofit secondary support chains, one set for each cylinder strap location.
Certifications: Stands are NFPA 1 and NFPA 55 approved, OSHA approved, and CGA-P1 approved.
Spécifications
- Catalog No:
- 76292-418
- 76292-416
- 76292-420
- 76292-414
- 76292-412
- 76292-422
- Cylinder type:
- Compressed gas
- Compressed gas
- Compressed gas
- Compressed gas
- Compressed gas
- Compressed gas
- Material:
- Steel
- Steel
- Steel
- Steel
- Steel
- Steel
- Mounting type:
- Floor
- Floor
- Floor
- Floor
- Floor
- Floor
- Straps:
- 2 per Cylinder
- 1 per Cylinder
- 2 per Cylinder
- 1 per Cylinder
- 2 per Cylinder
- 2 per Cylinder
- Strap type:
- Poly Cinch Strap with Steel Buckle
- Poly Cinch Strap with Steel Buckle
- Poly Cinch Strap with Steel Buckle
- Poly Cinch Strap with Steel Buckle
- Poly Cinch Strap with Steel Buckle
- Poly Cinch Strap with Steel Buckle