Gas Cylinder Forklift Pallets, Individually Secured, Justrite®
Fournisseur: Justrite
Certificats
A propos de cet article
This Individually Secured Gas Cylinder Forklift Pallet provides safety when moving and storing of gas cylinders.
- Fully enclosed 197 mm (7.75") wide forklift channels with two-way access
- Bumper guard edging with steel reinforced vinyl protects cylinders from damage, while providing extra grip
- Accepts cylinders from 102 to 305 mm (4" to 12") in diameter
- Secure transportation of multiple gas cylinders with a forklift using these pallets
Fully welded construction from 11 gauge (3 mm) hot-rolled steel, powder-coated to ensure longevity and chemical resistance.
Heavy-duty polypropylene support straps, 38×1372 mm (1.5×54") and rated at 516 kg (1138 lbs.), feature steel cinch buckle and hold individual cylinders securely against integral bracket.
For an improved level of safety for any installation, all brackets include openings to retrofit secondary support chains, one set for each cylinder strap location.
Certifications: Forklift pallets are NFPA 1 and NFPA 55 approved, OSHA approved, and CGA-P1 approved.
Spécifications
- Catalog No:
- 76292-358
- 76292-352
- 76292-356
- 76292-354
- Cylinder type:
- Compressed gas
- Compressed gas
- Compressed gas
- Compressed gas
- Material:
- Steel
- Steel
- Steel
- Steel
- Mounting type:
- Forklift
- Forklift
- Forklift
- Forklift
- Straps:
- 2 per Cylinder
- 2 per Cylinder
- 2 per Cylinder
- 2 per Cylinder
- Strap type:
- Poly Cinch Strap with Steel Buckle
- Poly Cinch Strap with Steel Buckle
- Poly Cinch Strap with Steel Buckle
- Poly Cinch Strap with Steel Buckle