Gas Cylinder Barricade Rack Pallet with Ramp, Justrite®
Fournisseur: Justrite
Certificats
A propos de cet article
These Gas Cylinder Barricade Rack Pallets with Ramp provide safety when moving and storing of gas cylinders.
- Secure cylinders up to 279 mm (11") in diameter
- Fully enclosed 197 mm (7.75") wide forklift channels with two-way access
- Secure transportation of multiple gas cylinders with a forklift using these end-load oriented barricade rack pallets
Barricade racks surround cylinders with structural steel tubing, continuously seam welded at all junction points and mounted to the pallet.
They are sealed with exterior grade powder-coat paint for long lasting strength, durability and dependability. Tops ground smooth and capped with high-density polyethylene (HDPE) caps to prevent corrosion and injuries.
Features two levels of welded steel chains for each cylinder and clasps to connect the chains to the rack, ensuring a snug fit.
Certifications: Rack pallets are NFPA 1 and NFPA 55 approved, OSHA approved, and CGA-P1 approved.
Spécifications
- Catalog No:
- 76292-350
- 76292-348
- Cylinder type:
- Compressed gas
- Compressed gas
- Material:
- Steel
- Steel
- Mounting type:
- Forklift
- Forklift
- Straps:
- 2 per Cylinder
- 2 per Cylinder
- Strap type:
- Chain
- Chain